siRNA Details
| Gene Symbol: Prkch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031085.2 | NP_112347.1 | 8 | 1222 | ||
| XM_008764720.1 | XP_008762942.1 | 8 | 1104 | |||
| GenBank mRNA | BC081782.1 | - | 8 | 1222 | 7123785 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkch| Gene Symbol: Prkch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031085.2 | NP_112347.1 | 8 | 1222 | ||
| XM_008764720.1 | XP_008762942.1 | 8 | 1104 | |||
| GenBank mRNA | BC081782.1 | - | 8 | 1222 | 7123785 | |