siRNA Details
| Gene Symbol: NEK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003157.4 | NP_003148.2 | 2 | 399 | ||
| XM_006713310.2 | XP_006713373.1 | 2 | 396 | |||
| XM_011534039.2 | XP_011532341.1 | 2 | 396 | |||
| XM_011534040.2 | XP_011532342.1 | 2 | 396 | |||
| XM_017007085.1 | XP_016862574.1 | 2 | 396 | |||
| XM_017007086.1 | XP_016862575.1 | 2 | 396 | |||
| GenBank mRNA | BC015515.1 | - | 2 | 363 | ||
| BC063044.1 | - | 2 | 389 | 5169184 | ||
| DB073127.1 | - | 2 | 399 | |||
| L20321.1 | - | 2 | 374 | |||

NEK4
Genomic Map