siRNA Details
| Gene Symbol: Ptger2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031088.1 | NP_112350.1 | 2 | 915 | ||
| GenBank mRNA | U48858.1 | - | 2 | 981 | ||
| U94708.1 | - | 2 | 915 | |||
More Information
Other Species Targeted
Set Membership: |
|

Ptger2
Genomic Map