siRNA Details
| Gene Symbol: Rab1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031090.2 | NP_112352.2 | 5 | 526 | ||
| GenBank mRNA | AY321327.1 | - | 482 | |||
| BC066662.1 | - | 5 | 526 | 5623442 | ||
| FQ213656.1 | - | 5 | 536 | |||
| FQ214472.1 | - | 5 | 536 | |||
| J02998.1 | - | 5 | 349 | |||

Rab1a
Genomic Map