siRNA Details
| Gene Symbol: Rab3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031091.2 | NP_112353.1 | 4 | 522 | ||
| XM_006238531.2 | XP_006238593.1 | 4 | 568 | |||
| GenBank mRNA | BC060562.1 | - | 4 | 440 | 6918181 | |
| U68184.1 | - | 3 | 228 | |||
| Y14019.1 | - | 3 | 410 | |||

Rab3b
Genomic Map