siRNA Details
| Gene Symbol: Rab13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031092.1 | NP_112354.1 | 3 | 294 | ||
| GenBank mRNA | AF525280.1 | - | 3 | 294 | ||
| FQ222701.1 | - | 3 | 220 | |||
| M83678.1 | - | 2 | 93 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab13| Gene Symbol: Rab13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031092.1 | NP_112354.1 | 3 | 294 | ||
| GenBank mRNA | AF525280.1 | - | 3 | 294 | ||
| FQ222701.1 | - | 3 | 220 | |||
| M83678.1 | - | 2 | 93 | |||