siRNA Details
Gene Symbol: Rala | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031093.2 | NP_112355.1 | 2 | 199 | ||
XM_006253993.2 | XP_006254055.1 | 3 | 403 | |||
XM_008771763.1 | XP_008769985.1 | 3 | 217 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rala | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031093.2 | NP_112355.1 | 2 | 199 | ||
XM_006253993.2 | XP_006254055.1 | 3 | 403 | |||
XM_008771763.1 | XP_008769985.1 | 3 | 217 |