siRNA Details
| Gene Symbol: Renbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031095.1 | NP_112357.1 | 8 | 944 | ||
| XM_006229615.2 | XP_006229677.1 | 9 | 1066 | |||
| GenBank mRNA | D10233.1 | - | 8 | 944 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Renbp| Gene Symbol: Renbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031095.1 | NP_112357.1 | 8 | 944 | ||
| XM_006229615.2 | XP_006229677.1 | 9 | 1066 | |||
| GenBank mRNA | D10233.1 | - | 8 | 944 | ||