siRNA Details
| Gene Symbol: Rpl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031100.1 | NP_112362.1 | 3 | 117 | ||
| GenBank mRNA | BC058467.1 | - | 3 | 106 | 6918973 | |
| FQ210278.1 | - | 3 | 101 | |||
| FQ218098.1 | - | 3 | 101 | |||
| FQ220555.1 | - | 3 | 100 | |||
| FQ221864.1 | - | 3 | 101 | |||
| FQ222021.1 | - | 3 | 101 | |||
| FQ229032.1 | - | 3 | 100 | |||
| FQ229696.1 | - | 1 | 228 | |||
| FQ233952.1 | - | 3 | 101 | |||
| X87106.1 | - | 3 | 117 | |||

Rpl10
Genomic Map