siRNA Details
| Gene Symbol: Rpl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031104.1 | NP_112366.1 | 5 | 352 | ||
| GenBank mRNA | BC058466.1 | - | 4 | 368 | 6887477 | |
| BC082750.1 | - | 4 | 343 | 7300771 | ||
| FQ222147.1 | - | 4 | 361 | |||
| FQ222660.1 | - | 4 | 361 | |||
| FQ222798.1 | - | 4 | 362 | |||
| FQ224693.1 | - | 4 | 360 | |||
| FQ228373.1 | - | 4 | 358 | |||
| X78444.1 | - | 5 | 352 | |||

Rpl22
Genomic Map