siRNA Details
| Gene Symbol: Rps6ka1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031107.1 | NP_112369.1 | 15, 16 | 1475 | ||
| GenBank mRNA | M99169.1 | - | 15, 16 | 1475 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rps6ka1| Gene Symbol: Rps6ka1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031107.1 | NP_112369.1 | 15, 16 | 1475 | ||
| GenBank mRNA | M99169.1 | - | 15, 16 | 1475 | ||