siRNA Details
| Gene Symbol: Rps27a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031113.1 | NP_112375.1 | 1 | 85 | ||
| Gene Symbol: LOC100912032 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751421.3 | XP_003751469.1 | 2, 3 | 149 | ||

Rps27a
Genomic Map