siRNA Details
| Gene Symbol: S100a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031114.1 | NP_112376.1 | 2, 3 | 172 | ||
| GenBank mRNA | AB453238.1 | - | 1, 2 | 127 | ||
| BC091565.1 | - | 2, 3 | 198 | 7441806 | ||
| FQ222734.1 | - | 2, 3 | 227 | |||
| FQ222778.1 | - | 2, 3 | 195 | |||
| FQ223502.1 | - | 2, 3 | 192 | |||
| FQ228469.1 | - | 2, 3 | 195 | |||
| FQ228745.1 | - | 2, 3 | 195 | |||
| FQ229216.1 | - | 2, 3 | 195 | |||
| J03627.1 | - | 2, 3 | 172 | |||

S100a10
Genomic Map