siRNA Details
| Gene Symbol: Sctr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031115.1 | NP_112377.1 | 5 | 629 | ||
| GenBank mRNA | BC081781.1 | - | 5 | 587 | 7189388 | |
| X59132.1 | - | 5 | 629 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sctr| Gene Symbol: Sctr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031115.1 | NP_112377.1 | 5 | 629 | ||
| GenBank mRNA | BC081781.1 | - | 5 | 587 | 7189388 | |
| X59132.1 | - | 5 | 629 | |||
Set Membership: |
|