siRNA Details
| Gene Symbol: Ssb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031119.2 | NP_112381.2 | 3 | 205 | ||
| XM_006234321.2 | XP_006234383.1 | 3 | 281 | |||
| XM_006234322.2 | XP_006234384.1 | 3 | 281 | |||
| XM_006234323.2 | XP_006234385.1 | 3 | 281 | |||
| GenBank mRNA | BC081780.1 | - | 3 | 205 | 7192454 | |
| BC097252.1 | - | 3 | 211 | |||
| X67859.1 | - | 2 | 140 | |||

Ssb
Genomic Map