siRNA Details
| Gene Symbol: St13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031122.1 | NP_112384.1 | 12 | 1126 | ||
| GenBank mRNA | BC078804.1 | - | 12 | 1133 | 7109473 | |
| FQ212434.1 | - | 1183 | ||||
| FQ228746.1 | - | 12 | 1096 | |||
| X82021.1 | - | 12 | 1126 | |||

St13
Genomic Map