siRNA Details
| Gene Symbol: Stxbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031126.1 | NP_112388.1 | 7 | 476 | ||
| GenBank mRNA | U20283.1 | - | 7 | 476 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stxbp2| Gene Symbol: Stxbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031126.1 | NP_112388.1 | 7 | 476 | ||
| GenBank mRNA | U20283.1 | - | 7 | 476 | ||