siRNA Details
| Gene Symbol: Tceb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031129.3 | NP_112391.1 | 3 | 195 | ||
| XM_008767596.1 | XP_008765818.1 | 2 | 201 | |||
| GenBank mRNA | BC058463.1 | - | 3 | 199 | 6921630 | |
| FQ212271.1 | - | 2 | 171 | |||
| L42855.1 | - | 2 | 155 | |||

Tceb2
Genomic Map