siRNA Details
Gene Symbol: Thpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031133.2 | NP_112395.1 | 959 | |||
XM_008768827.1 | XP_008767049.1 | 6 | 1921 | |||
XM_008768828.1 | XP_008767050.1 | 6 | 1279 | |||
XM_008768829.1 | XP_008767051.1 | 5 | 2257 | |||
GenBank mRNA | D32207.1 | - | 960 |
Gene Symbol: LOC103690065 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248552.2 | XP_006248614.1 | 6 | 2090 | ||
XM_006248553.2 | XP_006248615.1 | 6 | 2078 | |||
XM_008768815.1 | XP_008767037.1 | 6 | 1921 | |||
XM_008768816.1 | XP_008767038.1 | 6 | 1279 |
