siRNA Details
| Gene Symbol: Thra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017960.1 | NP_001017960.1 | 7 | 1062 | ||
| NM_031134.2 | NP_112396.2 | 6 | 930 | |||
| GenBank mRNA | CB728734.1 | - | 3 | 411 | ||
| M31174.1 | - | 1144 | ||||
| M31177.1 | - | 2 | 271 | |||
| X07409.1 | - | 5 | 591 | |||

Thra
Genomic Map