siRNA Details
| Gene Symbol: Thra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017960.1 | NP_001017960.1 | 5 | 703 | ||
| NM_031134.2 | NP_112396.2 | 4 | 571 | |||
| GenBank mRNA | CB580585.1 | - | 4 | 503 | ||
| CB728734.1 | - | 1 | 52 | |||
| M18028.1 | - | 5 | 593 | |||
| M31174.1 | - | 785 | ||||
| X07409.1 | - | 3 | 232 | |||
| X12744.1 | - | 4 | 235 | |||

Thra
Genomic Map