siRNA Details
| Gene Symbol: STK11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000455.4 | NP_000446.1 | 2 | 1430 | ||
| XM_005259617.2 | XP_005259674.1 | 2 | 940 | |||
| XM_005259618.3 | XP_005259675.1 | 2 | 940 | |||
| XM_011528209.1 | XP_011526511.1 | 3 | 1071 | |||
| GenBank mRNA | AB209553.1 | - | 2 | 405 | ||
| AF035625.1 | - | 2 | 653 | |||
| AF217978.1 | - | 3 | 1276 | |||
| AK314858.1 | - | 2 | 441 | |||
| BC007981.2 | - | 2 | 765 | 3689780 | ||
| BC019334.1 | - | 2 | 586 | 3946005 | ||
| EF644172.1 | - | 2 | 399 | |||
| GU573916.1 | - | 1 | 23 | |||
| KU178339.1 | - | 2 | 315 | |||
| U63333.1 | - | 2 | 315 | |||

STK11
Genomic Map