siRNA Details
| Gene Symbol: Cib1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031145.1 | NP_112407.1 | 6 | 573 | ||
| GenBank mRNA | AF136585.1 | - | 6 | 573 | ||
| BC091143.1 | - | 6 | 559 | 7308828 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cib1| Gene Symbol: Cib1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031145.1 | NP_112407.1 | 6 | 573 | ||
| GenBank mRNA | AF136585.1 | - | 6 | 573 | ||
| BC091143.1 | - | 6 | 559 | 7308828 | ||