siRNA Details
| Gene Symbol: Slc20a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031148.1 | NP_112410.1 | 9 | 1891 | ||
| XM_006234991.2 | XP_006235053.1 | 9 | 2115 | |||
| GenBank mRNA | AB000489.1 | - | 9 | 1931 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc20a1| Gene Symbol: Slc20a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031148.1 | NP_112410.1 | 9 | 1891 | ||
| XM_006234991.2 | XP_006235053.1 | 9 | 2115 | |||
| GenBank mRNA | AB000489.1 | - | 9 | 1931 | ||