siRNA Details
| Gene Symbol: Zp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031150.1 | NP_112412.1 | 10 | 992 | ||
| GenBank mRNA | AB000929.1 | - | 10 | 992 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zp2| Gene Symbol: Zp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031150.1 | NP_112412.1 | 10 | 992 | ||
| GenBank mRNA | AB000929.1 | - | 10 | 992 | ||