siRNA Details
| Gene Symbol: Mdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031151.2 | NP_112413.2 | 1, 2 | 177 | ||
| GenBank mRNA | BC063165.1 | - | 1, 2 | 177 | 6918633 | |
| DQ402951.1 | - | 11 | ||||
| FQ213817.1 | - | 1, 2 | 94 | |||
| FQ215385.1 | - | 1, 2 | 137 | |||
| FQ215647.1 | - | 1, 2 | 138 | |||
| FQ216782.1 | - | 1, 2 | 168 | |||
| FQ217232.1 | - | 1, 2 | 129 | |||
| FQ224847.1 | - | 1, 2 | 74 | |||
| FQ224979.1 | - | 1, 2 | 156 | |||
| X04240.1 | - | 1, 2 | 118 | |||

Mdh2
Genomic Map