siRNA Details
| Gene Symbol: Sharpin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031153.2 | NP_112415.1 | 2 | 575 | ||
| GenBank mRNA | AF203906.1 | - | 2 | 249 | ||
| BC083553.1 | - | 2 | 575 | 7189111 | ||
| FQ213170.1 | - | 2 | 193 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sharpin| Gene Symbol: Sharpin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031153.2 | NP_112415.1 | 2 | 575 | ||
| GenBank mRNA | AF203906.1 | - | 2 | 249 | ||
| BC083553.1 | - | 2 | 575 | 7189111 | ||
| FQ213170.1 | - | 2 | 193 | |||