siRNA Details
| Gene Symbol: Lrp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031322.3 | NP_112612.2 | 38 | 5526 | ||
| GenBank mRNA | AB011533.1 | - | 25 | 3692 | ||
| AB073317.3 | - | 38 | 5526 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrp4| Gene Symbol: Lrp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031322.3 | NP_112612.2 | 38 | 5526 | ||
| GenBank mRNA | AB011533.1 | - | 25 | 3692 | ||
| AB073317.3 | - | 38 | 5526 | |||