siRNA Details
| Gene Symbol: Bmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031323.1 | NP_112613.1 | 1 | 259 | ||
| XM_008770834.1 | XP_008769056.1 | 1 | 293 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bmp1| Gene Symbol: Bmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031323.1 | NP_112613.1 | 1 | 259 | ||
| XM_008770834.1 | XP_008769056.1 | 1 | 293 | |||