siRNA Details
| Gene Symbol: Cyr61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031327.2 | NP_112617.2 | 3 | 746 | ||
| GenBank mRNA | AB015877.1 | - | 3 | 611 | ||
| AF218568.1 | - | 3 | 746 | |||
| BC081689.1 | - | 3 | 726 | 7122117 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyr61| Gene Symbol: Cyr61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031327.2 | NP_112617.2 | 3 | 746 | ||
| GenBank mRNA | AB015877.1 | - | 3 | 611 | ||
| AF218568.1 | - | 3 | 746 | |||
| BC081689.1 | - | 3 | 726 | 7122117 | ||