siRNA Details
| Gene Symbol: Slc22a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031332.1 | NP_112622.1 | 2 | 445 | ||
| GenBank mRNA | AB017446.1 | - | 2 | 445 | ||
| BC081777.1 | - | 2 | 443 | 7109603 | ||
| FQ209755.1 | - | 457 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc22a8| Gene Symbol: Slc22a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031332.1 | NP_112622.1 | 2 | 445 | ||
| GenBank mRNA | AB017446.1 | - | 2 | 445 | ||
| BC081777.1 | - | 2 | 443 | 7109603 | ||
| FQ209755.1 | - | 457 | ||||