siRNA Details
Gene Symbol: Polr2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031335.3 | NP_112625.1 | 5 | 386 | ||
GenBank mRNA | AB017711.1 | - | 5 | 386 | ||
BC083552.1 | - | 5 | 426 | 7190444 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Polr2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031335.3 | NP_112625.1 | 5 | 386 | ||
GenBank mRNA | AB017711.1 | - | 5 | 386 | ||
BC083552.1 | - | 5 | 426 | 7190444 |