siRNA Details
| Gene Symbol: Kl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031336.1 | NP_112626.1 | 2 | 1231 | ||
| GenBank mRNA | AB017820.1 | - | 2 | 1231 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kl| Gene Symbol: Kl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031336.1 | NP_112626.1 | 2 | 1231 | ||
| GenBank mRNA | AB017820.1 | - | 2 | 1231 | ||