siRNA Details
| Gene Symbol: Parg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031339.1 | NP_112629.1 | 12, 13 | 2334 | ||
| XM_006252669.1 | XP_006252731.1 | 11, 12 | 2004 | |||
| GenBank mRNA | AB019366.1 | - | 12, 13 | 2334 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Parg| Gene Symbol: Parg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031339.1 | NP_112629.1 | 12, 13 | 2334 | ||
| XM_006252669.1 | XP_006252731.1 | 11, 12 | 2004 | |||
| GenBank mRNA | AB019366.1 | - | 12, 13 | 2334 | ||