siRNA Details
| Gene Symbol: Ppargc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031347.1 | NP_112637.1 | 12, 13 | 2336 | ||
| XM_008770219.1 | XP_008768441.1 | 12, 13 | 2403 | |||
| XM_008770220.1 | XP_008768442.1 | 11, 12 | 3573 | |||
| GenBank mRNA | AB025784.1 | - | 12, 13 | 2336 | ||
| AY237127.1 | - | 12, 13 | 2295 | |||

Ppargc1a
Genomic Map