siRNA Details
| Gene Symbol: Aplnr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031349.2 | NP_112639.1 | 1 | 719 | ||
| GenBank mRNA | AB033170.1 | - | 1 | 496 | ||
| AF090346.1 | - | 1 | 275 | |||
| AF184883.1 | - | 1 | 541 | |||
| BC072494.1 | - | 1 | 719 | 7097318 | ||
More Information
Set Membership: |
|

Aplnr
Genomic Map