siRNA Details
Gene Symbol: Aplnr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031349.2 | NP_112639.1 | 1 | 1079 | ||
GenBank mRNA | AB033170.1 | - | 1 | 856 | ||
AF090346.1 | - | 1 | 635 | |||
AF184883.1 | - | 1 | 901 | |||
BC072494.1 | - | 1 | 1079 | 7097318 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
