siRNA Details
| Gene Symbol: Rrad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053338.2 | NP_445790.2 | 3 | 590 | ||
| XM_006255055.2 | XP_006255117.1 | 3 | 672 | |||
| GenBank mRNA | CK602305.1 | - | 3 | 562 | ||
| U12187.1 | - | 3 | 562 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rrad| Gene Symbol: Rrad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053338.2 | NP_445790.2 | 3 | 590 | ||
| XM_006255055.2 | XP_006255117.1 | 3 | 672 | |||
| GenBank mRNA | CK602305.1 | - | 3 | 562 | ||
| U12187.1 | - | 3 | 562 | |||