siRNA Details
| Gene Symbol: Dbnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277211.1 | NP_001264140.1 | 9 | 872 | ||
| NM_001277212.1 | NP_001264141.1 | 9 | 860 | |||
| NM_001277213.1 | NP_001264142.1 | 9 | 857 | |||
| NM_031352.2 | NP_112642.1 | 9 | 863 | |||
| GenBank mRNA | AB009346.1 | - | 9 | 843 | ||
| AB038364.1 | - | 9 | 786 | |||
| AB038365.1 | - | 9 | 843 | |||
| AB039818.1 | - | 9 | 800 | |||
| AB039819.1 | - | 9 | 831 | |||
| BC072483.1 | - | 9 | 849 | 7104722 | ||
| FQ228854.1 | - | 9 | 860 | |||

Dbnl
Genomic Map