siRNA Details
| Gene Symbol: Vdac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031355.1 | NP_112645.1 | 4 | 230 | ||
| XM_006253358.2 | XP_006253420.1 | 5 | 233 | |||
| GenBank mRNA | AB039664.1 | - | 4 | 230 | ||
| AF048829.1 | - | 3 | 114 | |||
| AF048830.1 | - | 3 | 117 | |||
| AF268469.1 | - | 3 | 141 | |||
| BC061780.1 | - | 4 | 166 | 5623339 | ||
| FQ216638.1 | - | 5 | 218 | |||

Vdac3
Genomic Map