siRNA Details
| Gene Symbol: Mkln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031359.1 | NP_112649.1 | 11 | 1224 | ||
| GenBank mRNA | AB046442.1 | - | 11 | 1224 | ||
| FQ213447.1 | - | 1 | 65 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mkln1| Gene Symbol: Mkln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031359.1 | NP_112649.1 | 11 | 1224 | ||
| GenBank mRNA | AB046442.1 | - | 11 | 1224 | ||
| FQ213447.1 | - | 1 | 65 | |||