siRNA Details
| Gene Symbol: Smpd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031360.2 | NP_112650.1 | 10 | 1170 | ||
| XM_008773007.1 | XP_008771229.1 | 11 | 1107 | |||
| GenBank mRNA | AB047002.1 | - | 10 | 909 | ||
| BC091139.1 | - | 10 | 1170 | 7378100 | ||

Smpd2
Genomic Map