siRNA Details
Gene Symbol: Mchr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031758.1 | NP_113946.1 | 2 | 675 | ||
GenBank mRNA | AF008650.1 | - | 2 | 675 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
