siRNA Details
| Gene Symbol: Pafah1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031763.3 | NP_113951.1 | 1663 | |||
| GenBank mRNA | AF016049.1 | - | 1478 | |||
| BC072510.1 | - | 1663 | 7104733 | |||
| FQ224997.1 | - | 1388 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pafah1b1| Gene Symbol: Pafah1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031763.3 | NP_113951.1 | 1663 | |||
| GenBank mRNA | AF016049.1 | - | 1478 | |||
| BC072510.1 | - | 1663 | 7104733 | |||
| FQ224997.1 | - | 1388 | ||||