siRNA Details
| Gene Symbol: Cpz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031766.1 | NP_113954.1 | 11 | 1723 | ||
| XM_006251151.2 | XP_006251213.1 | 10 | 1718 | |||
| GenBank mRNA | AF017637.1 | - | 11 | 1723 | ||
| AF017638.1 | - | 11 | 1723 | |||

Cpz
Genomic Map