siRNA Details
| Gene Symbol: Pdxk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031769.1 | NP_113957.1 | 3 | 203 | ||
| GenBank mRNA | AF020346.1 | - | 3 | 203 | ||
| FQ213832.1 | - | 3 | 307 | |||
| Gene Symbol: RGD1566085 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772854.1 | XP_008771076.1 | 3 | 333 | ||

Pdxk
Genomic Map