siRNA Details
Gene Symbol: Thbd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031771.2 | NP_113959.1 | 1 | 1514 | ||
GenBank mRNA | AF022743.1 | - | 1 | 1439 | ||
BC070903.1 | - | 1 | 1514 | 7100705 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Thbd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031771.2 | NP_113959.1 | 1 | 1514 | ||
GenBank mRNA | AF022743.1 | - | 1 | 1439 | ||
BC070903.1 | - | 1 | 1514 | 7100705 |