siRNA Details
| Gene Symbol: Thbd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031771.2 | NP_113959.1 | 1 | 1468 | ||
| GenBank mRNA | AF022743.1 | - | 1 | 1393 | ||
| BC070903.1 | - | 1 | 1468 | 7100705 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thbd| Gene Symbol: Thbd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031771.2 | NP_113959.1 | 1 | 1468 | ||
| GenBank mRNA | AF022743.1 | - | 1 | 1393 | ||
| BC070903.1 | - | 1 | 1468 | 7100705 | ||