siRNA Details
| Gene Symbol: Polr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031773.1 | NP_113961.1 | 1 | 110 | ||
| GenBank mRNA | AF025424.1 | - | 1 | 110 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Polr1b| Gene Symbol: Polr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031773.1 | NP_113961.1 | 1 | 110 | ||
| GenBank mRNA | AF025424.1 | - | 1 | 110 | ||