siRNA Details
| Gene Symbol: Usf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031777.2 | NP_113965.1 | 8 | 717 | ||
| XM_006250266.2 | XP_006250328.1 | 8 | 902 | |||
| XM_008769765.1 | XP_008767987.1 | 6 | 553 | |||
| GenBank mRNA | AF026476.1 | - | 6 | 586 | ||
| BC088849.1 | - | 8 | 717 | 7313699 | ||

Usf1
Genomic Map